Få mere at vide
Pregabalin Methyl Ester Hydrochloride, Mikromol™
Beskrivelse
Pregabalin Methyl Ester Hydrochloride
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Pregabalin Methyl Ester Hydrochloride |
| CAS | 714230-22-5 |
| Analyt- eller komponentnavne | Pregabalin Methyl Ester Hydrochloride |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C9 H19 N O2 . Cl H |
| Certificeringer/compliance | ISO/IEC 17025 |
| Synonym | (S)-Pregabalin Methyl Ester Hydrochloride, Hexanoic acid, 3-(aminomethyl)-5-methyl-, methyl ester, hydrochloride (1:1), (3S)-, Hexanoic acid, 3-(aminomethyl)-5-methyl-, methyl ester, hydrochloride, (3S)- (9CI) |
| InChI formel | InChI=1S/C9H19NO2.ClH/c1-7(2)4-8(6-10)5-9(11)12-3;/h7-8H,4-6,10H2,1-3H3;1H/t8-;/m0./s1 |
| SMIL | Cl.COC(=O)C[C@@H](CN)CC(C)C |
| IUPAC navn | methyl (3S)-3-(aminomethyl)-5-methylhexanoate;hydrochloride |
| Vis mere |