Få mere at vide
Pregabalin, LoGiCal
Artikelnummer. 30462050
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30462050 10 mg
1 options
Artikelnummer. 30462050 Leverandør LoGiCal Leverandørnr. LGCFOR1376.00
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Pregabalin
CAS 148553-50-8
Analyt- eller komponentnavne Pregabalin
Forsendelsestilstand Room Temperature
Molekylær formel C8 H17 N O2
Certificeringer/compliance ISO 9001
Synonym Hexanoic acid, 3-(aminomethyl)-5-methyl-, (3S)-, Hexanoic acid, 3-(aminomethyl)-5-methyl-, (S)-, (2S)-3-(Aminomethyl)-5-methylhexanoic acid, (3S)-3-(Aminomethyl)-5-methylhexanoic acid, 3S-(aminomethyl)-5-methyl-hexanoic acid, (S)-(+)-3-(Aminomethyl)-5-methylhexanoic acid, (S)-3-(Ammoniomethyl)-5-methylhexanoate, (S)-Pregabalin, CI 1008, Gabanext 75, Lyrica, Mahagaba-75, Maxgalin 75, Neugaba 75, PD 144723, Pregabalin, Pregalin 75, (S)-Isobutylgaba
InChI formel InChI=1S/C8H17NO2/c1-6(2)3-7(5-9)4-8(10)11/h6-7H,3-5,9H2,1-2H3,(H,10,11)/t7-/m0/s1
Løsningstype Neat
SMIL CC(C)C[C@H](CN)CC(=O)O
IUPAC navn (3S)-3-(aminomethyl)-5-methylhexanoic acid
Industry Type Clinical Testing
Molekylvægt (g/mol) 159.23
Mængde 10 mg
Formel vægt 159.1259 g/mol