Få mere at vide
Uvex Preformed Mask silv-Air e 7312 FFP3
Shop All Uvex Products
Click to view available options
:
Filter Type: FFP3
Description
General Features
- Particle-filtering preformed mask with exhalation valve
- Color: white
Protection Features
- Certified FFP3 face mask to EN 149
- D label (dolomite dust test passed): maximum breathability even with really high dust levels
- R label (reusable): tested for re-usability and performance, including in the second shift
Comfort Features
- Soft material edges for increased wearer comfort
- Very soft, all-around sealing lip for optimum wearer comfort and secure fit
- Flexible, adjustable integrated nose clip
- Four-point headband attachment for a comfortable, individual and secure fit
- Exhalation valve to lessen exhalation resistance and to reduce heat and moisture inside the mask
Specifications
Specifications
| Certificeringer/overholdelse (Europa) | EN 149 (R marked, D marked) |
| Produkttype | Preformed Mask |
| Med udåndingsventil | Yes |
| Filtereffektivitet | FFP3 |
| Inkluderer | 360° exhalation valve, four-point head-securing strap, comfort nose clip, circumferential comfort sealing lip |