Få mere at vide
Prednisolone Tebutate, TRC
Artikelnummer. 30427540
Skift visning
Mængde:
100 mg
200 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30427540 100 mg
30412019 200 mg
30418576 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30427540 Leverandør Toronto Research Chemicals Leverandørnr. TRCP703910100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 7681-14-3
Molekylær formel C27H38O6
Molekylvægt (g/mol) 458.59
Synonym 11beta,17,21-Trihydroxypregna-1,4-diene-3,20-dione 21-(3,3-Dimethylbutyrate), 3,3-Dimethylbutyric Acid 21-Ester with 11beta,17,21-Trihydroxypregna-1,4-diene-3,20-dione, 11beta,17,21-Trihydroxypregna-1,4-diene-3,20-dione 21-(3,3-dimethylbutyrate), Codelcortone TBA, Hydeltra TBA, Predalone TBA, Prednisolone 21-tert-Butylacetate, Prednisolone TBA, Prednisolone Tebutate, Prednisolone tert-Butylacetate
IUPAC navn [2-[(8S,9S,10R,11S,13S,14S,17R)-11,17-dihydroxy-10,13-dimethyl-3-oxo-7,8,9,11,12,14,15,16-octahydro-6H-cyclopenta[a]phenanthren-17-yl]-2-oxoethyl] 3,3-dimethylbutanoate
SMIL CC(C)(C)CC(=O)OCC(=O)[C@@]1(O)CC[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@H]3[C@@H](O)C[C@]12C
Tekniske data
Kemisk navn eller materiale Prednisolone Tebutate
Mængde 100 mg
InChI formel InChI=1S/C27H38O6/c1-24(2,3)14-22(31)33-15-21(30)27(32)11-9-19-18-7-6-16-12-17(28)8-10-25(16,4)23(18)20(29)13-26(19,27)5/h8,10,12,18-20,23,29,32H,6-7,9,11,13-15H2,1-5H3/t18-,19-,20-,23+,25-,26-,27-/m0/s1
Formel vægt 458.2668