Få mere at vide
Prednisolone Acetate, LoGiCal
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Prednisolone Acetate |
| CAS | 52-21-1 |
| Analyt- eller komponentnavne | Prednisolone Acetate |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C23 H30 O6 |
| Certificeringer/compliance | ISO 9001 |
| Synonym | Hydrocortisone Acetate Imp. C (EP), Methylprednisolone Acetate Imp. E (EP), Prednisolone Imp. C (EP), Prednisolone Acetate, 11beta,17-Dihydroxy-3,20-dioxo-pregna-1,4-dien-21-yl Acetate, Hydrocortisone Acetate Imp. C (Pharmeuropa), (11beta)-21-(Acetyloxy)-11,17-dihydroxypregna-1,4-diene-3,20-dione, 11beta,17alpha,21-Trihydroxypregna-1,4-diene-3,20-dione 21-acetate, 21-(Acetoxy)-11beta,17alpha-dihydroxypregna-1,4-diene-3,20-dione, 21-Acetoxy-11beta,17-dihydroxypregna-1,4-diene-3,20-dione, Ak-Tate, Cormalone, Cortipred, Deltilen, Econopred, Falcon, Falcon (steroid), Hydroprednisone acetate, Inflanefran, Inflanefran Forte, Meticortelone acetate, Meticotelone acetate, NSC 10966, Nisolone, Pred Mild, Pred-Forte, Predalone 50, Prediacortin, Predicort, Prednelan N, Prednidoren, Prednisolone 21-acetate, Prednisolone acetate, Prenema, Supercortyl, Hydrocortisone Acetate Impurity C, Methylprednisolone Acetate Impurity E, Prednisolone Impurity C, Prednisone Impurity L |
| InChI formel | InChI=1S/C23H30O6/c1-13(24)29-12-19(27)23(28)9-7-17-16-5-4-14-10-15(25)6-8-21(14,2)20(16)18(26)11-22(17,23)3/h6,8,10,16-18,20,26,28H,4-5,7,9,11-12H2,1-3H3/t16-,17-,18-,20+,21-,22-,23-/m0/s1 |
| Løsningstype | Neat |
| SMIL | CC(=O)OCC(=O)[C@@]1(O)CC[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@H]3[C@@H](O)C[C@]12C |
| Vis mere |