Få mere at vide
Prednisolone 21-Ethylcarbonate, TRC
Artikelnummer. 30135029
Skift visning
Mængde:
250 mg
2.5 g
Pakningsstørrelse:
2.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30135029 2.5 g 2.5g
17934335 250 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30135029 Leverandør Toronto Research Chemicals Leverandørnr. TRCP7037352.5G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 2205-88-1
Molekylær formel C24 H32 O7
Molekylvægt (g/mol) 432.51
Synonym Pregna-1,4-diene-3,20-dione, 21-[(ethoxycarbonyl)oxy]-11,17-dihydroxy-, (11β)-, Pregna-1,4-diene-3,20-dione, 11β,17,21-trihydroxy-, 21-(ethyl carbonate) (6CI,8CI), (11β)-21-[(Ethoxycarbonyl)oxy]-11,17-dihydroxypregna-1,4-diene-3,20-dione, 11β,17-Dihydroxy-3,20-dioxopregna-1,4-dien-21-yl Ethyl Carbonate, Prednisolone 21-Ethylcarbonate, Prednicarbate Imp. D (EP)
IUPAC navn [2-[(8S,9S,10R,11S,13S,14S,17R)-11,17-dihydroxy-10,13-dimethyl-3-oxo-7,8,9,11,12,14,15,16-octahydro-6H-cyclopenta[a]phenanthren-17-yl]-2-oxoethyl] ethyl carbonate
SMIL CCOC(=O)OCC(=O)[C@@]1(O)CC[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@H]3[C@@H](O)C[C@]12C
Tekniske data
Kemisk navn eller materiale Prednisolone 21-Ethylcarbonate
Mængde 2.5 g
InChI formel InChI=1S/C24H32O7/c1-4-30-21(28)31-13-19(27)24(29)10-8-17-16-6-5-14-11-15(25)7-9-22(14,2)20(16)18(26)12-23(17,24)3/h7,9,11,16-18,20,26,29H,4-6,8,10,12-13H2,1-3H3/t16-,17-,18-,20+,22-,23-,24-/m0/s1
Formel vægt 432.2148