Få mere at vide
Prednisolone 21-Acetate 17-Valerate, TRC
Artikelnummer. 17983935
Skift visning
Mængde:
250 mg
1 g
2.5 g
Pakningsstørrelse:
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
17983935 2.5 g 2.5g
17961865 250 mg -
17921335 1 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 17983935 Leverandør Toronto Research Chemicals Leverandørnr. TRCP7038302.5G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 72064-79-0
Molekylær formel C28 H38 O7
Molekylvægt (g/mol) 486.6
Synonym (11β)-21-(Acetyloxy)-11-hydroxy-17-[(1-oxopentyl)oxy]pregna-1,4-diene-3,20-dione, Pregna-1,4-diene-3,20-dione, 11β,17,21-trihydroxy-, 21-acetate 17-valerate (7CI), Acepreval, Lidomex, Prednisolone 17-valerate 21-acetate, Prednisolone 21-acetate 17-valerate, Prednival 21-acetate
IUPAC navn [(8S,9S,10R,11S,13S,14S,17R)-17-(2-acetyloxyacetyl)-11-hydroxy-10,13-dimethyl-3-oxo-7,8,9,11,12,14,15,16-octahydro-6H-cyclopenta[a]phenanthren-17-yl] pentanoate
SMIL CCCCC(=O)O[C@@]1(CC[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@H]3[C@@H](O)C[C@]12C)C(=O)COC(=O)C
Tekniske data
Kemisk navn eller materiale Prednival Acetate
Renhedsgrad noter HPLC
Mængde 2.5 g
InChI formel InChI=1S/C28H38O7/c1-5-6-7-24(33)35-28(23(32)16-34-17(2)29)13-11-21-20-9-8-18-14-19(30)10-12-26(18,3)25(20)22(31)15-27(21,28)4/h10,12,14,20-22,25,31H,5-9,11,13,15-16H2,1-4H3/t20-,21-,22-,25+,26-,27-,28-/m0/s1
Formel vægt 486.2618
Procent renhed >95%