missing translation for 'onlineSavingsMsg'
Få mere at vide

Prednisolone 17-Acetate, TRC

Product Code. 30427562
Change view
Click to view available options
Mængde:
25 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30427562 25 mg
30417405 100 mg
30418166 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30427562 Supplier Toronto Research Chemicals Supplier No. TRCP70383525MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52628-64-5
Molekylær formel C23 H30 O6
Molekylvægt (g/mol) 402.48
Synonym Prednisolone 17-Acetate, (11β)-17-(Acetyloxy)-11,21-dihydroxypregna-1,4-diene-3,20-dione
IUPAC navn [(8S,9S,10R,11S,13S,14S,17R)-11-hydroxy-17-(2-hydroxyacetyl)-10,13-dimethyl-3-oxo-7,8,9,11,12,14,15,16-octahydro-6H-cyclopenta[a]phenanthren-17-yl] acetate
SMIL CC(=O)O[C@@]1(CC[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@H]3[C@@H](O)C[C@]12C)C(=O)CO

Specifications

Kemisk navn eller materiale Prednisolone 17-Acetate
Mængde 25 mg
InChI formel InChI=1S/C23H30O6/c1-13(25)29-23(19(28)12-24)9-7-17-16-5-4-14-10-15(26)6-8-21(14,2)20(16)18(27)11-22(17,23)3/h6,8,10,16-18,20,24,27H,4-5,7,9,11-12H2,1-3H3/t16-,17-,18-,20+,21-,22-,23-/m0/s1
Formel vægt 402.2042
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.