Få mere at vide
Pre-Vitamin D3 Octanoate (>80%), TRC
Artikelnummer. 30426930
Skift visning
Mængde:
0.5 mg
Pakningsstørrelse:
0.5mg
Artikelnummer. Mængde unitSize
30426930 0.5 mg 0.5mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30426930 Leverandør Toronto Research Chemicals Leverandørnr. TRCP8221700.5MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Pre-Vitamin D3 Octanoate
CAS 927822-17-1
Molekylær formel C35H58O2
Mængde 0.5 mg
Synonym (1S)-3-[(1Z)-2-[(1R,3aR,7aR)-1-[(1R)-1,5-Dimethylhexyl]-2,3,3a,6,7,7a-hexahydro-7a-methyl-1H-inden-4-yl]ethenyl]-4-methyl-3-cyclohexen-1-yl Octanoate, (1S)-3-[(1Z)-2-[(1R,3aR,7aR)-1-[(1R)-1,5-dimethylhexyl]-2,3,3a,6,7,7a-hexahydro-7a-methyl-1H-inden-4-yl]ethenyl]-4-methyl-3-cyclohexen-1-yl ester Octanoic acid
InChI formel InChI=1S/C35H58O2/c1-7-8-9-10-11-17-34(36)37-31-21-18-27(4)30(25-31)20-19-29-16-13-24-35(6)32(22-23-33(29)35)28(5)15-12-14-26(2)3/h16,19-20,26,28,31-33H,7-15,17-18,21-25H2,1-6H3/b20-19-/t28-,31+,32-,33+,35-/m1/s1
SMIL CCCCCCCC(=O)O[C@H]1CCC(=C(C1)\C=C/C2=CCC[C@]3(C)[C@H](CC[C@@H]23)[C@H](C)CCCC(C)C)C
IUPAC navn [(1S)-3-[(Z)-2-[(1R,3aR,7aR)-7a-methyl-1-[(2R)-6-methylheptan-2-yl]-1,2,3,3a,6,7-hexahydroinden-4-yl]ethenyl]-4-methylcyclohex-3-en-1-yl] octanoate
Molekylvægt (g/mol) 510.85
Formel vægt 510.4437