Learn More
Keystone Cleanroom Products Pre-Sterilized Sharpie, Fine Point
Product Code. 31008264 Shop All Keystone Cleanroom Products Products
Change view
Click to view available options
Color:
Black
Blue
Unit Size:
Pack of 250
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
31008264 Black Pack of 250
31008262 Blue Pack of 250
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 31008264 Supplier Keystone Cleanroom Products Supplier No. S10FMARKBLK
  • ONLINE EXCLUSIVE PRICE
    2965.00 DKK
    2377.89 DKK / Pakke med 250
    Spare 587.11 DKK
    19% Slukket
Forventet forsendelse: 11-01-2027
Ready-to-use sterile permanent ink Sharpie markers eliminate the risks associated with surface sanitization procedures.
  • Gamma Irradiated Sterile to a 10-6 SAL
  • Lot controlled
  • Ready to use
  • Packaged in easy to open Tyvek™ peel pouches
  • Minimize contamination risks
  • Simplifies conformance to SOP's
  • Eliminate the need for wiping down contaminated writing instruments
Specifications
Material Plastic
Ruled No
Color Black
Cover Color Black
Autoclavable Not Autoclavable
Certifications/Compliance Certificate of Sterility
Disposable Limited-use
Packaging 10 Pk.
Product Type Sterile Marker
Sterility Sterile
Tip Style Fine Point