Få mere at vide
Novus Biologicals™ pre-mRNA cleavage factor I (59 kDa subunit) Overexpression Lysate
Shop alle Bio Techne produkterBeskrivelse
The pre-mRNA cleavage factor I (59 kDa subunit) Overexpression Lysate is derived from human. It has been validated for the following applications: Western Blot.
Tekniske data
Tekniske data
| Cellelinje | The lysate was created in HEK293T cells, using plasmid ID RC22127 and based on accession number NM_24811. The protein contains a C-terminal DDK tag. |
| Til brug med (applikation) | Western Blot |
| Mængde | 0.1 mg |
| Opbevaringsbuffer | RIPA buffer |
| Renhed | Protein |
| Testspecificitet | Homo sapiens pre-mRNA cleavage factor I, 59 kDa subunit (FLJ12529), transcript variant 1, mRNA. |
| Indhold og opbevaring | -80°C. Avoid freeze-thaw cycles. |
| Proteintilstand | Native |
For Research Use Only