Få mere at vide
Saint Gobain Life Sciences Pre-drilled Rubber Stopper Without Holes
Beskrivelse
- Designed to keep container contents secure
- Ideal for securing glass, metal and plastic containers
- Easy to use
Tekniske data
Tekniske data
| Højde (metrisk) | 30mm |
| Mængde | 50/Pk. |
| Farve | Blå |
| Diameter (metrisk) bund | 23 mm |
| Diameter (metrisk) Top | 29 mm |
| Materiale | Natural Rubber |
| Produkttype | Rubber Stopper |