Få mere at vide
Prazobind-d8, TRC
Artikelnummer. 30409976
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30409976 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30409976 Leverandør Toronto Research Chemicals Leverandørnr. TRCP7021521MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Prazobind-d8
CAS 1189701-23-2
Molekylær formel C23H19D8N5O3
Mængde 1 mg
InChI formel InChI=1S/C23H27N5O3/c1-30-19-12-17-18(13-20(19)31-2)25-23(26-21(17)24)28-9-7-27(8-10-28)22(29)16-11-14-3-5-15(16)6-4-14/h3,5,11-15H,4,6-10H2,1-2H3,(H2,24,25,26)/i7D2,8D2,9D2,10D2
SMIL [2H]C1([2H])N(C(=O)C2=CC3CCC2C=C3)C([2H])([2H])C([2H])([2H])N(c4nc(N)c5cc(OC)c(OC)cc5n4)C1([2H])[2H]
IUPAC navn [4-(4-amino-6,7-dimethoxyquinazolin-2-yl)-2,2,3,3,5,5,6,6-octadeuteriopiperazin-1-yl]-(3-bicyclo[2.2.2]octa-2,5-dienyl)methanone
Molekylvægt (g/mol) 429.54
Formel vægt 429.2616