Få mere at vide
Prazitone Hydrochloride, LoGiCal
Artikelnummer. 30461919
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30461919 10 mg
1 options
Artikelnummer. 30461919 Leverandør LoGiCal Leverandørnr. LGCFOR3229.01
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Prazitone Hydrochloride
CAS 20432-72-8
Analyt- eller komponentnavne Prazitone Hydrochloride
Forsendelsestilstand Room Temperature
Molekylær formel C16 H19 N3 O3 . Cl H
Certificeringer/compliance ISO 9001
Synonym 5-Phenyl-5-(2-piperidinylmethyl)-2,4,6(1H,3H,5H)-pyrimidinetrione hydrochloride, 5-Phenyl-5-(2-piperidylmethyl)barbituric acid hydrochloride, AGN 511, Prazitone hydrochloride
InChI formel InChI=1S/C16H19N3O3.ClH/c20-13-16(11-6-2-1-3-7-11,14(21)19-15(22)18-13)10-12-8-4-5-9-17-12;/h1-3,6-7,12,17H,4-5,8-10H2,(H2,18,19,20,21,22);1H
Løsningstype Neat
SMIL Cl.O=C1NC(=O)C(CC2CCCCN2)(C(=O)N1)c3ccccc3
IUPAC navn 5-phenyl-5-(piperidin-2-ylmethyl)-1,3-diazinane-2,4,6-trione;hydrochloride
Industry Type Clinical Testing
Molekylvægt (g/mol) 337.8
Mængde 10 mg
Formel vægt 337.1193 g/mol