Få mere at vide
Praziquantel-d11, TRC
Artikelnummer. 30422027
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30422027 25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30422027 Leverandør Toronto Research Chemicals Leverandørnr. TRCP70209725MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Praziquantel cyclohexyl-d11
CAS 1246343-36-1
Renhedsgrad noter HPLC
Molekylær formel C19 2H11 H13 N2 O2
Mængde 25 mg
Synonym 4H-Pyrazino[2,1-a]isoquinolin-4-one','2-(cyclohexyl-1,2,2,3,3,4,4,5,5,6,6-d11-carbonyl)-1,2,3,6,7,11b-hexahydro-, 2-(Cyclohexyl-1,2,2,3,3,4,4,5,5,6,6-d11-carbonyl)-1,2,3,6,7,11b-hexahydro-4H-pyrazino[2,1-a]isoquinolin-4-one, Praziquantel D11 (cyclohexyl D11)
InChI formel InChI=1S/C19H24N2O2/c22-18-13-20(19(23)15-7-2-1-3-8-15)12-17-16-9-5-4-6-14(16)10-11-21(17)18/h4-6,9,15,17H,1-3,7-8,10-13H2/i1D2,2D2,3D2,7D2,8D2,15D
SMIL [2H]C1([2H])C([2H])([2H])C([2H])([2H])C([2H])(C(=O)N2CC3N(CCc4ccccc34)C(=O)C2)C([2H])([2H])C1([2H])[2H]
IUPAC navn 2-(1,2,2,3,3,4,4,5,5,6,6-undecadeuteriocyclohexanecarbonyl)-3,6,7,11b-tetrahydro-1H-pyrazino[2,1-a]isoquinolin-4-one
Molekylvægt (g/mol) 323.47
Formel vægt 323.2528
Procent renhed >95