Få mere at vide
Pravastatin Lactone-d3, TRC
Artikelnummer. 30410964
Skift visning
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30410964 1 mg
30409682 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30410964 Leverandør Toronto Research Chemicals Leverandørnr. TRCP7011921MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1217769-04-4
Molekylær formel C23 2H3 H31 O6
Molekylvægt (g/mol) 409.53
Synonym (2S)-2-(Methyl-d3)butanoic acid (1S,3S,7S,8S,8aR)-1,2,3,7,8,8a-hexahydro-3-hydroxy-7-methyl-8-[2-[(2R,4R)-tetrahydro-4-hydroxy-6-oxo-2H-pyran-2-yl]ethyl]-1-naphthalenyl ester, (+)-Pravastatin lactone-d3, Pravastatin lactone-d3, R 414-d3, SQ 31369-d3, (1S,3S,7S,8S,8aR)-3-Hydroxy-8-[2-[(2R,4R)-4-hydroxy-6-oxotetrahydro-2H-pyran-2-yl]ethyl]-7-methyl-1,2,3,7,8,8a-hexahydronaphthalen-1-yl (2S)-2-(methyl-d3)butanoate, Pravastatin lactone D3 (2-(Methyl-D3)butanoic acid)
IUPAC navn [(1S,3S,7S,8S,8aR)-3-hydroxy-8-[2-[(2R,4R)-4-hydroxy-6-oxooxan-2-yl]ethyl]-7-methyl-1,2,3,7,8,8a-hexahydronaphthalen-1-yl] (2S)-2-(trideuteriomethyl)butanoate
SMIL [2H]C([2H])([2H])[C@@H](CC)C(=O)O[C@H]1C[C@H](O)C=C2C=C[C@H](C)[C@H](CC[C@@H]3C[C@@H](O)CC(=O)O3)[C@@H]12
Tekniske data
Kemisk navn eller materiale Pravastatin Lactone methyl-d3
Mængde 1 mg
InChI formel InChI=1S/C23H34O6/c1-4-13(2)23(27)29-20-11-16(24)9-15-6-5-14(3)19(22(15)20)8-7-18-10-17(25)12-21(26)28-18/h5-6,9,13-14,16-20,22,24-25H,4,7-8,10-12H2,1-3H3/t13-,14-,16+,17+,18+,19-,20-,22-/m0/s1/i2D3
Formel vægt 409.2544