missing translation for 'onlineSavingsMsg'
Få mere at vide

Pravastatin Ethyl Ester, Mikromol™

Artikelnummer. 30049482
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30049482 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30049482 Leverandør Mikromol Leverandørnr. MM0503.100025

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Pravastatin Ethyl Ester
TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale Pravastatin Ethyl Ester
CAS 2727065-87-2
Analyt- eller komponentnavne Pravastatin Ethyl Ester
Forsendelsestilstand Room Temperature
Molekylær formel C25 H40 O7
Certificeringer/compliance ISO 9001
Synonym Pravastatin Ethyl Ester
InChI formel InChI=1S/C25H40O7/c1-5-15(3)25(30)32-22-13-19(27)11-17-8-7-16(4)21(24(17)22)10-9-18(26)12-20(28)14-23(29)31-6-2/h7-8,11,15-16,18-22,24,26-28H,5-6,9-10,12-14H2,1-4H3/t15-,16-,18+,19+,20+,21-,22-,24-/m0/s1
SMIL CCOC(=O)C[C@H](O)C[C@H](O)CC[C@H]1[C@@H](C)C=CC2=C[C@@H](O)C[C@H](OC(=O)[C@@H](C)CC)[C@H]12
IUPAC navn ethyl (3R,5R)-7-[(1S,2S,6S,8S,8aR)-6-hydroxy-2-methyl-8-[(2S)-2-methylbutanoyl]oxy-1,2,6,7,8,8a-hexahydronaphthalen-1-yl]-3,5-dihydroxyheptanoate
Molekylvægt (g/mol) 452.58
Mængde 25 mg
Formel vægt 452.2774 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.