Få mere at vide
Prasugrel Thiolactone-d5 (Mixture of Diastereomers), TRC
Artikelnummer. 30424250
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30424250 5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30424250 Leverandør Toronto Research Chemicals Leverandørnr. TRCP7011675MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Prasugrel Thiolactone-d5 (Mixture of Diastereomers)
CAS 2124271-74-3
Molekylær formel C18H13D5FNO2S
Mængde 5 mg
Synonym 5-[2-(Cyclopropyl-d5)-1-(2-fluorophenyl)-2-oxoethyl]-5,6,7,7a-tetrahydrothieno[3,2-c]pyridin-2(4H)-one, R 95913-d5, 2-Oxo-Prasugrel-d4
InChI formel InChI=1S/C18H18FNO2S/c19-14-4-2-1-3-13(14)17(18(22)11-5-6-11)20-8-7-15-12(10-20)9-16(21)23-15/h1-4,9,11,15,17H,5-8,10H2/i5D2,6D2,11D
SMIL FC1=CC=CC=C1C(C(C2([2H])C([2H])([2H])C2([2H])[2H])=O)N3CCC4C(C3)=CC(S4)=O
IUPAC navn 5-(2-(cyclopropyl-d5)-1-(2-fluorophenyl)-2-oxoethyl)-5,6,7,7a-tetrahydrothieno[3,2-c]pyridin-2(4H)-one
Molekylvægt (g/mol) 336.44
Formel vægt 336.14