Få mere at vide
Prasugrel EP G as Hydrochloride, Mikromol™
Artikelnummer. 17902526
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
17902526 25 mg
1 options
Artikelnummer. 17902526 Leverandør Mikromol Leverandørnr. MM3637.010025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Prasugrel EP G as Hydrochloride
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1-Cyclopropyl-2-(2-fluorophenyl)ethane-1,2-dione
CAS 1391054-37-7
Analyt- eller komponentnavne 1-Cyclopropyl-2-(2-fluorophenyl)ethane-1,2-dione
Forsendelsestilstand Ice pack (-20°C)
Molekylær formel C11 H9 F O2
Certificeringer/compliance ISO 9001
Synonym 1-Cyclopropyl-2-(2-fluorophenyl)ethane-1,2-dione, Prasugrel Hydrochloride Impurity G as Hydrochloride, Prasugrel Impurity G as Hydrochloride
InChI formel InChI=1S/C11H9FO2/c12-9-4-2-1-3-8(9)11(14)10(13)7-5-6-7/h1-4,7H,5-6H2
SMIL Fc1ccccc1C(=O)C(=O)C2CC2
IUPAC navn 1-cyclopropyl-2-(2-fluorophenyl)ethane-1,2-dione
Molekylvægt (g/mol) 192.19
Mængde 25 mg
Formel vægt 192.0587 g/mol