Få mere at vide
Prasugrel-d5, TRC
Artikelnummer. 30412421
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30412421 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30412421 Leverandør Toronto Research Chemicals Leverandørnr. TRCP7011521MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Prasugrel-d5
CAS 1127252-92-9
Molekylær formel C20H15D5FNO3S
Mængde 1 mg
Synonym 2-[2-(Acetyloxy)-6,7-dihydrothieno[3,2-c]pyridin-5(4H)-yl]-1-(cyclopropyl-d5)-2-(2-fluorophenyl)ethanone, CS 747-d5, Effient-d5, LY 640315-d5
InChI formel InChI=1S/C20H20FNO3S/c1-12(23)25-18-10-14-11-22(9-8-17(14)26-18)19(20(24)13-6-7-13)15-4-2-3-5-16(15)21/h2-5,10,13,19H,6-9,11H2,1H3/i6D2,7D2,13D
SMIL FC1=CC=CC=C1C(C(C2([2H])C([2H])([2H])C2([2H])[2H])=O)N3CCC(SC(OC(C)=O)=C4)=C4C3
Molekylvægt (g/mol) 378.47
Formel vægt 378.1462