Få mere at vide
Prasterone Acetate, Mikromol™
Beskrivelse
Prasterone Acetate
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Dehydroisoandrosterone Acetate (DHEA Acetate) |
| CAS | 853-23-6 |
| Analyt- eller komponentnavne | Dehydroisoandrosterone Acetate (DHEA Acetate) |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C21 H30 O3 |
| Certificeringer/compliance | ISO 17025 |
| Synonym | Androstenolone acetate, DHEA acetate, Dehydroepiandrost-5-en-17-one 3-acetate, Dehydroepiandrosterone 3-acetate, Dehydroisoandrosterone 3-acetate, Dehydroisoandrosterone acetate, Prasterone acetate, SKF 2847, trans-Dehydroandrosterone acetate, Androst-5-en-17-one, 3-(acetyloxy)-, (3beta)-, Androst-5-en-17-one, 3beta-hydroxy-, acetate (6CI,8CI), 17-Oxoandrost-5-en-3beta-yl acetate, 3beta-Acetoxy-5-androstene-17-one, 3beta-Acetoxyandrost-5-en-17-one, 3beta-Acetoxydehydroepiandrosterone, 3beta-Acetyloxyandrost-5-en-17-one, 3beta-Hydroxy-5-androsten-17-one acetate, 3beta-Hydroxy-5-androstene-17-one acetate, 3beta-Hydroxyandrost-5-en-17-one 3-acetate, 5-Androsten-3beta-ol-17-one acetate, Delta5-Dehydroepiandrosterone 3-acetate |
| InChI formel | InChI=1S/C21H30O3/c1-13(22)24-15-8-10-20(2)14(12-15)4-5-16-17-6-7-19(23)21(17,3)11-9-18(16)20/h4,15-18H,5-12H2,1-3H3/t15-,16-,17-,18-,20-,21-/m0/s1 |
| SMIL | CC(=O)O[C@H]1CC[C@]2(C)[C@H]3CC[C@@]4(C)[C@@H](CCC4=O)[C@@H]3CC=C2C1 |
| IUPAC navn | [(3S,8R,9S,10R,13S,14S)-10,13-dimethyl-17-oxo-1,2,3,4,7,8,9,11,12,14,15,16-dodecahydrocyclopenta[a]phenanthren-3-yl] acetate |
| Vis mere |