Få mere at vide
PRAK Antibody (OTI5E1), PerCP, Novus Biologicals™
Artikelnummer. 30028280 Shop alle Bio Techne produkter
Skift visning
Mængde:
0.1 mL
Artikelnummer. Mængde
30028280 0.1 mL
1 options
Artikelnummer. 30028280 Leverandør Novus Biologicals Leverandørnr. NBP273594PCP
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
Mouse Monoclonal Antibody
PRAK Monoclonal antibody specifically detects PRAK in Human, Mouse, Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Tekniske data
Antigen PRAK
Applikationer Western Blot
Klassifikation Monoclonal
Klon OTI5E1
Konjugeret PerCP
Fortynding Western Blot
Formulering PBS
Gene Alias EC 2.7.11, MAP kinase-activated protein kinase 5, MAPK-activated protein kinase 5, MAPKAP kinase 5, MAPKAPK-5, mitogen-activated protein kinase-activated protein kinase 5, p38-regulated/activated protein kinase, PRAKEC 2.7.11.1
Værtsarter Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 197-471 of human MAPKAPK5 (NP_003659) produced in E.coli.
Oprensningsmetode Immunogen affinity purified
Mængde 0.1 mL
Regulatorisk status RUO
Forskningsdisciplin Protein Kinase
Primær eller sekundær Primary
Gen-id (Entrez) 8550
Målarter Human, Mouse, Rat
Indhold og opbevaring Store at 4C in the dark.
Form Purified
Isotype IgG2a