Få mere at vide
PPQ-102, MedChemExpress
Artikelnummer. 16476406
Encompass_Preferred
Skift visning
Mængde:
5 mg
10 mg
50 mg
100 mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
16476406 100 mg
16452127 50 mg
16408607 5 mg
16405988 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Artikelnummer. 16476406 Leverandør Medchem Express Leverandørnr. HY14179
MedChemExpress PPQ-102 is a potent CFTR inhibitor which can completely inhibited CFTR chloride current with IC50 of ∼90 nM.
PPQ-102 is a potent CFTR inhibitor which can completely inhibited CFTR chloride current with IC50 of ∼90 nM.
Tekniske data
Kemisk navn eller materiale PPQ-102
CAS 931706-15-9
Farve Light Yellow
Renhedsgrad noter Research
Molekylær formel C26H22N4O3
Mængde 100 mg
Synonym CFTR Inhibitor
Opløselighedsinformation DMSO : 50 mg/mL (114.03 mM; Need ultrasonic)
SMIL CN1C(C2=C(C3=CC=CC=C3)N4C5=CC=CC=C5NC(C6=CC=C(C)O6)C4=C2N(C)C1=O)=O
Molekylvægt (g/mol) 438.48
Formel vægt 438.48
Procent renhed 99.82%
Grad Research
Fysisk form Solid