Få mere at vide
PPP2R1B Antibody [Alexa Fluor« 594], Novus Biologicals Biologicals™
Artikelnummer. 30498836 Shop alle Bio Techne produkter
Skift visning
Mængde:
0.1 mL
Pakningsstørrelse:
0.1ml
Artikelnummer. Mængde unitSize
30498836 0.1 mL 0.1ml
1 options
Artikelnummer. 30498836 Leverandør Novus Biologicals Leverandørnr. NBP337951AF594
Rabbit Polyclonal Antibody
PPP2R1B Polyclonal antibody specifically detects PPP2R1B in Human samples. It is validated for ELISA,Western Blot,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Tekniske data
Antigen PPP2R1B
Applikationer ELISA, Western Blot, Immunocytochemistry/Immunofluorescence
Klassifikation Polyclonal
Konjugeret Alexa Fluor 594
Formulering 50mM Sodium Borate
Gene Alias MGC26454, PP2A subunit A isoform PR65-beta, PP2A subunit A isoform R1-beta, PP2A, subunit A, PR65-beta isoform, PP2A, subunit A, R1-beta isoform, PP2A-Abeta, PR65B, protein phosphatase 2 (formerly 2A), regulatory subunit A (PR 65), beta isoform, protein phosphatase 2 (formerly 2A), regulatory subunit A, beta isoform, protein phosphatase 2, regulatory subunit A, beta, protein phosphatase 2, structural/regulatory subunit A, beta, serine/threonine-protein phosphatase 2A 65 kDa regulatory subunit A betaisoform
Værtsarter Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 468-667 of human PPP2R1B (NP_859050.1).,, Sequence:, YAIREAATNNLMKLVQKFGTEWAQNTIVPKVLVMANDPNYLHRMTTLFCINALSEACGQEITTKQMLPIVLKMAGDQVANVRFNVAKSLQKIGPILDTNALQGEVKPVLQKLGQDEDMDVKYFAQEAISVVAQRLRKLEFPVKDSGEPSVPRADKNHFPRPTVPGEDMGKGPVYQLRGDTRDTLAQLGIAELVHFSQSTD
Oprensningsmetode Affinity purified
Mængde 0.1 mL
Regulatorisk status RUO
Forskningsdisciplin Breast Cancer, Protein Phosphatase, Wnt Signaling Pathway
Primær eller sekundær Primary
Gen-id (Entrez) 5519
Målarter Human
Indhold og opbevaring Store at 4°C in the dark.
Produkttype Antibody
Form Purified
Isotype IgG