missing translation for 'onlineSavingsMsg'
Få mere at vide

Invitrogen™ POU3F3 Monoclonal Antibody (CL15302)
GREENER_CHOICE

Artikelnummer. 30827053
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30827053 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30827053 Leverandør Invitrogen™ Leverandørnr. MA563019

Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!

Denne vare kan ikke returneres. Se returpolicy

Mouse Monoclonal Antibody

Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.

POU3F3 is a member of the class III POU family of transcription factors that are expressed in the central nervous system. The POU domain in these proteins is required for high affinity binding to octamer DNA sequences.
TRUSTED_SUSTAINABILITY

Specifikationer

Antigen POU3F3
Applikationer Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Klassifikation Monoclonal
Klon CL15302
Koncentration 1.0 mg/mL
Konjugeret Unconjugated
Formulering PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene POU3F3
Genadgangsnr. P20264, P31361, Q63262
Gene Alias 8-Oct; Brain-1; brain-specific homeobox/POU domain protein 1; Brn1; Brn-1; brn-1 protein; class III POU protein POU-domain; oct-8; Octamer-binding protein 8; octamer-binding transcription factor 8; Otf8; OTF-8; POU class 3 homeobox 3; POU domain, class 3, transcription factor 3; Pou3f3; RHS1; Rhs2; Skin1
Gensymboler POU3F3
Værtsarter Mouse
Immunogen Recombinant protein corresponding to Human POU3F3.
Oprensningsmetode Protein A
Mængde 100 μL
Regulatorisk status RUO
Primær eller sekundær Primary
Gen-id (Entrez) 18993, 192109, 5455
Målarter Human, Mouse, Rat
Indhold og opbevaring Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Produkttype Antibody
Form Liquid
Isotype IgG1
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.