Learn More
Memmert™ Potential-Free Contact (24 V / 2 A) with Socket to NAMUR NE 28 for Combined Error Message for UN160
Potential-free contact for combination error message (e.g. supply failure, sensor fault, fuse).
Brand:  Memmert™ UN160H6
Product Code. 17569885
  • 2200.00 DKK / Each
Only null left
Estimated Shipment: 25-07-2025
Description
  • Accessory
  • For combination error message
Specifications
Specifications
UN160 Oven
Socket to NAMUR NE 28
2 A
Potential-free Contact
24 V
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Memmert™ Potential-Free Contact (24 V / 2 A) with Socket to NAMUR NE 28 for Combined Error Message for UN160 > For Use With: UN160