Learn More
Potassiummethanesulfonate, 98%, BLD Pharm™
Product Code. 30947467
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
Unit Size:
100g
1g
25g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30947467 1 g 1g
30945917 25 g 25g
30943344 100 g 100g
30938442 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 30947467 Supplier BLD Pharm Supplier No. BD1488351g
 Restricted Item
Forventet forsendelse: 27-11-2026
Specifications
CAS 2386-56-3
Molecular Formula CH3KO3S
MDL Number MFCD00070544
Quantity 1 g
SMILES CS(=O)([O-])=O.[K+]
Molecular Weight (g/mol) 134.20
Percent Purity 98%
Chemical Name or Material Potassiummethanesulfonate