missing translation for 'onlineSavingsMsg'
Learn More

Potassium thiophene-3-trifluoroborate, 97%, Thermo Scientific™

Catalog Number p-7052689
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
192863-37-9
C4H3BF3KS
190.032
MFCD03701626
ULOOPHJJEPUNNH-UHFFFAOYSA-N
potassium 3-thiophenetrifluoroborate, potassium thiophene-3-trifluoroborate, potassium trifluoro thiophen-3-yl boranuide, potassium trifluoro thiophen-3-yl borate, pubchem11540, amtb173, potassiurn 3-thiophenetri tluoroborate, potassiotrifluoro 3-thienyl boron v, potassium trifluoro 3-thiophenyl boranuide, potassium trifluoro thiophen-3-yl borate 1-
23677379
potassium;trifluoro(thiophen-3-yl)boranuide
[B-](C1=CSC=C1)(F)(F)F.[K+]
This item is not returnable. View return policy
192863-37-9
C4H3BF3KS
190.032
MFCD03701626
ULOOPHJJEPUNNH-UHFFFAOYSA-N
potassium 3-thiophenetrifluoroborate, potassium thiophene-3-trifluoroborate, potassium trifluoro thiophen-3-yl boranuide, potassium trifluoro thiophen-3-yl borate, pubchem11540, amtb173, potassiurn 3-thiophenetri tluoroborate, potassiotrifluoro 3-thienyl boron v, potassium trifluoro 3-thiophenyl boranuide, potassium trifluoro thiophen-3-yl borate 1-
23677379
potassium;trifluoro(thiophen-3-yl)boranuide
[B-](C1=CSC=C1)(F)(F)F.[K+]

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Color White
Physical Form Powder
Quantity 250 mg
Formula Weight 190.04
Percent Purity 97%
Chemical Name or Material Potassium thiophene-3-trifluoroborate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'