missing translation for 'onlineSavingsMsg'
Learn More

Potassium thiocyanate, 98%, Thermo Scientific Chemicals

Catalog Number p-7021411
Click to view available options
Quantity:
500 g
2500 g
333-20-0
CKNS
97.176
MFCD00011413
ZNNZYHKDIALBAK-UHFFFAOYSA-M
potassium thiocyanate, potassium rhodanate, potassium rhodanide, rodanca, potassium sulfocyanate, rhodanide, rhocya, potassium thiocyanide, kscn, thiocyanic acid, potassium salt
516872
CHEBI:30951
potassium;thiocyanate
C(#N)[S-].[K+]
333-20-0
CKNS
97.176
MFCD00011413
ZNNZYHKDIALBAK-UHFFFAOYSA-M
potassium thiocyanate, potassium rhodanate, potassium rhodanide, rodanca, potassium sulfocyanate, rhodanide, rhocya, potassium thiocyanide, kscn, thiocyanic acid, potassium salt
516872
CHEBI:30951
potassium;thiocyanate
C(#N)[S-].[K+]

Potassium thiocyanate is used in the preparation of ethylenesulfide, lead thiocyante, which is used in the conversion of acyl chloride to thiocyanates. It is also used in solid state NMR of heavy-metal nuclei which results in the preparation of several mercury-thiocyanate and selenocyanate compounds as potential models. It finds application in cosmetic and personal care product formulations as a skin conditioning agent. It is also used in moisturizers and oral care products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Potassium thiocyanate is used in the preparation of ethylenesulfide, lead thiocyante, which is used in the conversion of acyl chloride to thiocyanates. It is also used in solid state NMR of heavy-metal nuclei which results in the preparation of several mercury-thiocyanate and selenocyanate compounds as potential models. It finds application in cosmetic and personal care product formulations as a skin conditioning agent. It is also used in moisturizers and oral care products.

Solubility
Soluble in water, acetone and alcohol.

Notes
Hygroscopic. Air, light and moisture sensitive. Incompatible with nitric acids, active halogen compounds and strong bases.
TRUSTED_SUSTAINABILITY
Melting Point 172°C to 175°C
Density 1.89
pH 5.3 to 8.5
Boiling Point 500°C (decomposition)
Odor Odorless
Quantity 500 g
Beilstein 3594799
Sensitivity Hygroscopic; Light sensitive
Merck Index 14,7691
Solubility Information Soluble in water,acetone and alcohol.
Formula Weight 97.18
Percent Purity 98%
Chemical Name or Material Potassium thiocyanate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'