Få mere at vide
Potassium Thiocyanate-13C, TRC
Artikelnummer. 30133528
Skift visning
Mængde:
10 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30133528 50 mg
30135405 10 mg
30005877 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30133528 Leverandør Toronto Research Chemicals Leverandørnr. TRCP69805850MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 143827-33-2
Molekylær formel 13CKNS
Molekylvægt (g/mol) 98.17
SMIL N#[13C]S[K]
Tekniske data
Kemisk navn eller materiale Potassium Thiocyanate-13C
Mængde 50 mg
InChI formel InChI=1S/CHNS.K/c2-1-3;/h3H;/q;+1/p-1/i1+1;
Formel vægt 97.9422