Få mere at vide
Potassium Thiocyanate-13C,15N, TRC
Artikelnummer. 30133052
Skift visning
Mængde:
5 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30133052 50 mg
30010940 25 mg
30009959 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30133052 Leverandør Toronto Research Chemicals Leverandørnr. TRCP69852250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 143673-61-4
Molekylær formel 13C 15N S . K
Molekylvægt (g/mol) 99.17
Synonym Potassium Thiocyanate-13C,15N, Thiocyanic-13C-15N acid, potassium salt (9CI)
IUPAC navn potassium;(^{15}N)azanylidyne(1^{13}C)methanethiolate
SMIL [K+].[S-][13C]#[15N]
Tekniske data
Kemisk navn eller materiale Potassium Thiocyanate-13C,15N
Mængde 50 mg
InChI formel InChI=1S/CHNS.K/c2-1-3;/h3H;/q;+1/p-1/i1+1,2+1;
Formel vægt 98.9392