Få mere at vide
Potassium Tetradecanoate-2,2-d2, CDN
Artikelnummer. 30318355
Skift visning
Mængde:
0.5 g
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30318355 0.5 g
30316688 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30318355 Leverandør CDN Leverandørnr. CDND51680.5G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Potassium Tetradecanoate-2,2-D2
CAS 352438-85-8
Molekylær formel C14 D2 H25 O2 . K
Mængde 0.5 g
Synonym Tetradecanoic-2,2-d2 acid, potassium salt (9 CI), Tetradecanoic-2,2-d2 acid potassium salt, Myristic acid-d2 potassium salt, Myristic-2,2-d2 acid potassium salt, Potassium tetradecanoate-2,2-D2
InChI formel InChI=1 S/C14H28O2.K/c1-2-3-4-5-6-7-8-9-10-11-12-13-14(15)16;/h2-13H2,1H3,(H,15,16);/q;+1/p-1/i13D2;
SMIL [K+].[2 H]C([2 H])(CCCCCCCCCCCC)C(=O)[O-]
IUPAC navn potassium;2,2-dideuteriotetradecanoate
Molekylvægt (g/mol) 268.474
Formel vægt 268.177 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity