Få mere at vide
Kaliumtetrachloroplatinum(II), TRC
Specifications
Specifications
| Kemisk navn eller materiale | Potassium Tetrachloroplatinum(II) |
| CAS | 10025-99-7 |
| Molekylær formel | Cl4 Pt . 2 K |
| Mængde | 5 g |
| Synonym | Platinate(2-)','tetrachloro-','potassium (1:2)','(SP-4-1)-, Platinate(2-)','tetrachloro-','dipotassium (8CI), Platinate(2-)','tetrachloro-','dipotassium','(SP-4-1)- (9CI), Potassium chloroplatinate(II) (6CI), Dipotassium platinum tetrachloride, Dipotassium tetrachloroplatinate, Dipotassium tetrachloroplatinate(2-), Dipotassium tetrachloroplatinate(II), NSC 175496, Platinous potassium chloride, Platinum potassium chloride (PtK2Cl4), Potassium chloroplatinate, Potassium chloroplatinate (K2PtCl4), Potassium platinochloride, Potassium platinous chloride, Potassium platinum(II) tetrachloride, Potassium tetrachloroplatinate, Potassium tetrachloroplatinate K2PtCl4, Potassium tetrachloroplatinate(2-), Potassium tetrachloroplatinate(II), Potassium tetrachloroplatinum(II), Tetrachloroplatinum(II) acid potassium salt |
| InChI formel | InChI=1S/4ClH.2K.Pt/h4*1H;;;/q;;;;2*+1;+2/p-4 |
| SMIL | [K+].[K+].[Cl-][Pt+2]([Cl-])([Cl-])[Cl-] |
| Molekylvægt (g/mol) | 415.09 |
| Formel vægt | 412.7676 |