Learn More
Potassium Sulfate, Extra Pure, SLR, Fisher Chemical™
Brand: Fisher Chemical P/6960/53
Additional Details : CAS Number : 7778-80-5 Weight : 0.50000kg
Quantity | 500g |
---|
Chemical Identifiers
7778-80-5 | |
174.253 | |
OTYBMLCTZGSZBG-UHFFFAOYSA-L | |
24507 | |
dipotassium;sulfate |
K2O4S | |
11388 | |
potassium sulfate, dipotassium sulfate, potassium sulphate, sulfuric acid dipotassium salt, sal polychrestum, sulfuric acid, potassium salt, arcanum duplicatum, kalium sulphuricum, caswell no. 702, sulfuric acid, dipotassium salt | |
CHEBI:32036 | |
[O-]S(=O)(=O)[O-].[K+].[K+] |
Specifications
7778-80-5 | |
11388 | |
OTYBMLCTZGSZBG-UHFFFAOYSA-L | |
dipotassium;sulfate | |
24507 | |
174.25g/mol | |
Plastic powder jar | |
White | |
6 to 8 |
K2O4S | |
potassium sulfate, dipotassium sulfate, potassium sulphate, sulfuric acid dipotassium salt, sal polychrestum, sulfuric acid, potassium salt, arcanum duplicatum, kalium sulphuricum, caswell no. 702, sulfuric acid, dipotassium salt | |
[O-]S(=O)(=O)[O-].[K+].[K+] | |
174.253 | |
CHEBI:32036 | |
Solid | |
1689°C | |
1067°C | |
500g |