missing translation for 'onlineSavingsMsg'
Learn More

Potassium Sulfate, Extra Pure, SLR, Fisher Chemical™

Catalog Number p-7073450
Click to view available options
Quantity:
500 g
1 kg
5 kg
This item is not returnable. View return policy
7778-80-5
K2O4S
174.25
MFCD00011388
OTYBMLCTZGSZBG-UHFFFAOYSA-L
potassium sulfate, dipotassium sulfate, potassium sulphate, sulfuric acid dipotassium salt, sal polychrestum, sulfuric acid, potassium salt, arcanum duplicatum, kalium sulphuricum, caswell no. 702, sulfuric acid, dipotassium salt
24507
CHEBI:32036
[K+].[K+].[O-]S([O-])(=O)=O
This item is not returnable. View return policy
7778-80-5
K2O4S
174.25
MFCD00011388
OTYBMLCTZGSZBG-UHFFFAOYSA-L
potassium sulfate, dipotassium sulfate, potassium sulphate, sulfuric acid dipotassium salt, sal polychrestum, sulfuric acid, potassium salt, arcanum duplicatum, kalium sulphuricum, caswell no. 702, sulfuric acid, dipotassium salt
24507
CHEBI:32036
[K+].[K+].[O-]S([O-])(=O)=O
Boiling Point 1689°C
Melting Point 1067°C
Color White
pH 6 to 8
Physical Form Solid
Quantity 500 g
IUPAC Name dipotassium;sulfate
Formula Weight 174.25g/mol
Packaging Plastic powder jar
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'