Få mere at vide
Potassium propionate, TRC
Artikelnummer. 30427274
Skift visning
Mængde:
5 g
10 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30427274 5 g
30412699 50 g
30418934 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30427274 Leverandør Toronto Research Chemicals Leverandørnr. TRCP6952935G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 327-62-8
Molekylær formel C3H5KO2
Molekylvægt (g/mol) 112.17
Synonym Propanoic acid, potassium salt (9CI), Propionic acid, potassium salt (8CI), Potassium propanoate, Potassium propionate
IUPAC navn potassium;propanoate
SMIL [K+].CCC(=O)[O-]
Tekniske data
Kemisk navn eller materiale Potassium propionate
Mængde 5 g
InChI formel InChI=1S/C3H6O2.K/c1-2-3(4)5;/h2H2,1H3,(H,4,5);/q;+1/p-1
Formel vægt 111.9927