missing translation for 'onlineSavingsMsg'
Learn More

Potassium permanganate, Puriss., meets analytical spec. of BP, Ph. Eur., USP, 99 to 100.5%, Honeywell Fluka™
BIOPHARMA

Catalog Number 15641280
Click to view available options
Quantity:
250 g
500 g
1 kg
2.5 kg
25 kg
6 x 1 kg
This item is not returnable. View return policy
Regulatory: This product contains a category 2 drug precursor according to Regulation (EC) No 273/2004, all customers must complete a declaration before delivery.
7722-64-7
KMnO4
158.032
MFCD00011364
VZJVWSHVAAUDKD-UHFFFAOYSA-N
potassium permanganate, chameleon mineral, condy's crystals, argucide, cairox, permanganate of potash, insta-perm, walko tablets, algae-k, solo san soo
516875
potassium;permanganate
[O-][Mn](=O)(=O)=O.[K+]
This item is not returnable. View return policy
Regulatory: This product contains a category 2 drug precursor according to Regulation (EC) No 273/2004, all customers must complete a declaration before delivery.
7722-64-7
KMnO4
158.032
MFCD00011364
VZJVWSHVAAUDKD-UHFFFAOYSA-N
potassium permanganate, chameleon mineral, condy's crystals, argucide, cairox, permanganate of potash, insta-perm, walko tablets, algae-k, solo san soo
516875
potassium;permanganate
[O-][Mn](=O)(=O)=O.[K+]

Puriss., meets analytical specification of BP, Ph. Eur., USP, 99-100.5%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY
CAS Max % 100.0000%
Quantity 25 kg
Assay Percent Range 99 to 100.5%
Linear Formula KMnO4
UN Number UN1490
Formula Weight 158.03
Percent Purity 99 to 100.5%
Grade puriss.
Packaging Paper / Poly Bag
Density 2.700 g/cm3
Chemical Name or Material Potassium permanganate, Puriss., meets analytical specification of BP, Ph. Eur., USP, 99-100.5%
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'