missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Permanganate, Certified AR for Analysis, meets analytical specification of Ph.Eur., BP, USP, Fisher Chemical™
Click to view available options
Quantity:
250 g
500 g
5 kg
Packaging:
Glass Jar
Plastic keg
Specifications
Melting Point | 240°C |
Color | Brown |
pH | 8 |
Physical Form | Powder/Solid |
Quantity | 250 g |
Formula Weight | 158.04g/mol |
Packaging | Glass Jar |
Safety and Handling
Product Identifier
- Potassium permanganate
- Danger
- Oxidising solid Category 2
- Acute toxicity Category 4
- Skin corrosion/irritation Category 1 C
- Serious eye damage/eye irritation Category 1
- Specific target organ toxicity after repeated exposure Category 2
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
- H272-May intensify fire; oxidiser.
- H302-Harmful if swallowed.
- H314-Causes severe skin burns and eye damage.
- H373-May cause damage to organs.
- H410-Very toxic to aquatic life with long lasting effects.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'