Få mere at vide
Kaliumpermanganat, certificeret AR til analyse, opfylder analytiske specifikationer fra Ph.Eur., BP, USP , Fisher Chemical™
Artikelnummer. 10063163 Shop alle Fisher Chemical produkter
Mængde:
250 g
500 g
5 kg
Emballage:
Glaskrukke
Plastfad
Pakningsstørrelse:
250g
500g
5kg
Regulatory: This product contains a category 2 drug precursor according to Regulation (EC) No 273/2004, all customers must complete a declaration before delivery.
Artikelnummer. 10063163
Brand: Fisher Chemical P/6560/65
Request Bulk or Custom Format
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
Regulatory: This product contains a category 2 drug precursor according to Regulation (EC) No 273/2004, all customers must complete a declaration before delivery.
Kemiske identifikatorer
CAS 7722-64-7
Molekylær formel KMnO4
Molekylvægt (g/mol) 158.032
MDL nummer 11364
InChI nøgle VZJVWSHVAAUDKD-UHFFFAOYSA-N
Synonym potassium permanganate, chameleon mineral, condy's crystals, argucide, cairox, permanganate of potash, insta-perm, walko tablets, algae-k, solo san soo
PubChem CID 516875
IUPAC navn kalium; permanganat
SMIL [O-][Mn](=O)(=O)=O.[K+]
Tekniske data
Smeltepunkt 240°C
Farve Brun
pH 8
Fysisk form Pulver/faststof
Mængde 5 kg
Formel vægt 158.04g/mol
Emballage Plastfad