missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium oxalate monohydrate, 99%, ACS reagent, Thermo Scientific Chemicals
349.27 DKK valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 356.0°C |
Color | White |
Infrared Spectrum | Authentic |
Assay Percent Range | 99% |
Packaging | Plastic bottle |
Linear Formula | KO2CCO2K·H2O |
Quantity | 500 g |
Merck Index | 15, 7772 |
Solubility Information | Solubility in water: 364g/L (20°C) |
Formula Weight | 184.24 |
Show More |
Safety and Handling
Product Identifier
- Potassium oxalate monohydrate
- Warning
- Acute toxicity Category 4
- H312-Harmful in contact with skin.
- H302-Harmful if swallowed.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'