Learn More
Potassium oxalate monohydrate, ACS, 98.5-101.0%, Thermo Scientific Chemicals
Description
Potassium oxalate monohydrate is used as a chelating agent; a purifying agent in pharmaceutical industry and a precipitating agent in rare-earth metal processing. It finds application as a grinding agent, a rust remover for metal treatment, a bleaching agent in textile and wood industry. It is also used in waste water treatment and as a scale remover in automobile radiators. Furthermore, it is used as a reagent in analytical chemistry for the determination of calcium and other heavy metals as well as a reducing agent in photography. It is also involved in the preparation of SBA-15 (nanostructured SiO2) supported vanadium oxide catalyst.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | >300°C |
Color | White |
Density | 2.13 g/mL |
pH | 6 to 8.5 |
Odor | Odorless |
Linear Formula | K2C2O4·H2O |
Quantity | 100 g |
Beilstein | 3752576 |
Merck Index | 14,7651 |
Solubility Information | Soluble in water. |
Show More |
RUO – Research Use Only