Learn More
Potassium nitrate, ReagentPlus™, Honeywell™
Catalog Number p-7072308
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
7757-79-1
KNO3
101.10
MFCD00011409
FGIUAXJPYTZDNR-UHFFFAOYSA-N
potassium nitrate, saltpeter, nitre, nitrate of potash, vicknite, nitric acid potassium salt, nitric acid, potassium salt, saltpetre, kaliumnitrat, kalii nitras
24434
CHEBI:63043
potassium;nitrate
[K+].[O-][N+]([O-])=O
7757-79-1
KNO3
101.10
MFCD00011409
FGIUAXJPYTZDNR-UHFFFAOYSA-N
potassium nitrate, saltpeter, nitre, nitrate of potash, vicknite, nitric acid potassium salt, nitric acid, potassium salt, saltpetre, kaliumnitrat, kalii nitras
24434
CHEBI:63043
potassium;nitrate
[K+].[O-][N+]([O-])=O
Reagent Grade,≥99.0%
  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY
Quantity 500 g
UN Number UN1486
Formula Weight 101.1g/mol
Percent Purity ≥99%
Grade ReagentPlus™
Chemical Name or Material Potassium nitrate