missing translation for 'onlineSavingsMsg'
Learn More

Potassium Nitrate, ≥99.0%, ACS Reagent, Honeywell Fluka™

Catalog Number 15642110
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Poly Bottle
This item is not returnable. View return policy
7757-79-1
KNO3
101.10
MFCD00011409
FGIUAXJPYTZDNR-UHFFFAOYSA-N
potassium nitrate, saltpeter, nitre, nitrate of potash, vicknite, nitric acid potassium salt, nitric acid, potassium salt, saltpetre, kaliumnitrat, kalii nitras
24434
CHEBI:63043
[K+].[O-][N+]([O-])=O
This item is not returnable. View return policy
7757-79-1
KNO3
101.10
MFCD00011409
FGIUAXJPYTZDNR-UHFFFAOYSA-N
potassium nitrate, saltpeter, nitre, nitrate of potash, vicknite, nitric acid potassium salt, nitric acid, potassium salt, saltpetre, kaliumnitrat, kalii nitras
24434
CHEBI:63043
[K+].[O-][N+]([O-])=O

ACS Reagent,≥99.0%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY
Quantity 500 g
UN Number UN1486
IUPAC Name potassium;nitrate
Formula Weight 101.1g/mol
Percent Purity ≥99.0%
Grade ACS Reagent
Packaging Poly Bottle
Chemical Name or Material Potassium Nitrate
compliance-icons
Product Identifier
  • Potassium nitrate
Signal Word
  • Warning
Hazard Category
  • Oxidising solid Category 3
Hazard Statement
  • H272-May intensify fire; oxidiser.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'