missing translation for 'onlineSavingsMsg'
Learn More

Potassium Iodate, ACS Reagent, 99.5%, Honeywell™ Fluka™

Catalog Number p-7120714
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
12 kg
4 x 2.5 kg
Packaging:
Drum with Inliner
Poly Bottle
This item is not returnable. View return policy
7758-05-6
IKO3
214.00
MFCD00011406
JLKDVMWYMMLWTI-UHFFFAOYSA-M
potassium iodate, potassium triodate, caswell no. 693a, iodic acid, potassium salt, unii-i139e44nhl, potassium iodine oxide kio3, iodic acid hio3 , potassium salt, epa pesticide chemical code 075703, iodic acid hio3 , potassium salt 1:1, kaliumjodat
23665710
potassium;iodate
[O-]I(=O)=O.[K+]
This item is not returnable. View return policy
7758-05-6
IKO3
214.00
MFCD00011406
JLKDVMWYMMLWTI-UHFFFAOYSA-M
potassium iodate, potassium triodate, caswell no. 693a, iodic acid, potassium salt, unii-i139e44nhl, potassium iodine oxide kio3, iodic acid hio3 , potassium salt, epa pesticide chemical code 075703, iodic acid hio3 , potassium salt 1:1, kaliumjodat
23665710
potassium;iodate
[O-]I(=O)=O.[K+]

ACS Reagent, 99.5%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY
Melting Point 560°C
Color Colorless
pH 5 to 8 (at 20°C)
Physical Form Solid
Quantity 5 g
Linear Formula KIO3
UN Number UN1479
Formula Weight 214g/mol
Percent Purity 99.50%
Grade ACS Reagent
Packaging Poly Bottle
Appearance Crystals
Density 3.93g/mL at 25°C; Bulk Density: ca. 2400kg/cu.m
Chemical Name or Material Potassium Iodate
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'