missing translation for 'onlineSavingsMsg'
Learn More

Potassium hydroxide concentrate, For 1L standard solution, 0.1 M KOH (0.1N), Honeywell Fluka™

Catalog Number p-7122101
336.38 DKK valid until 2024-03-31
Use promo code "21663" to get your promotional price.


Click to view available options
Quantity:
1 L
6 x 1 Ea.
1310-58-3
HKO
56.11
MFCD00003553
KWYUFKZDYYNOTN-UHFFFAOYSA-M
potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
14797
CHEBI:32035
potassium;hydroxide
[OH-].[K+]
1310-58-3
HKO
56.11
MFCD00003553
KWYUFKZDYYNOTN-UHFFFAOYSA-M
potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
14797
CHEBI:32035
potassium;hydroxide
[OH-].[K+]

for 1L standard solution, 0.1 M KOH (0.1N)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY
Linear Formula KOH
UN Number UN1814
Quantity 1 L
Formula Weight 56.11g/mol
Packaging Ampoule
Chemical Name or Material Potassium hydroxide concentrate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'