missing translation for 'onlineSavingsMsg'
Learn More

Potassium hydrogen sulfate, 97%, Thermo Scientific Chemicals

Catalog Number p-7082091
Click to view available options
Quantity:
10,000 g
100 g
2500 g
500 g
This item is not returnable. View return policy
7646-93-7
HKO4S
136.16
MFCD00011404
CHKVPAROMQMJNQ-UHFFFAOYSA-M
potassium bisulfate, potassium hydrogen sulfate, potassium bisulphate, sulfuric acid, monopotassium salt, potassium acid sulfate, sal enixum, potassium hydrogensulphate, monopotassium sulfate, acid potassium sulfate, caswell no. 682c
516920
potassium;hydrogen sulfate
[K+].OS([O-])(=O)=O
This item is not returnable. View return policy
7646-93-7
HKO4S
136.16
MFCD00011404
CHKVPAROMQMJNQ-UHFFFAOYSA-M
potassium bisulfate, potassium hydrogen sulfate, potassium bisulphate, sulfuric acid, monopotassium salt, potassium acid sulfate, sal enixum, potassium hydrogensulphate, monopotassium sulfate, acid potassium sulfate, caswell no. 682c
516920
potassium;hydrogen sulfate
[K+].OS([O-])(=O)=O

Potassium hydrogen sulfate is used in the conversion of tartrates to bitartrates in wine. It serves as a disintegrating agent in analytical chemistry and as an intermediate in the preparation of potassium persulfate by electrolysis. It is also employed in the manufacturing of fertilizers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Potassium hydrogen sulfate is used in the conversion of tartrates to bitartrates in wine. It serves as a disintegrating agent in analytical chemistry and as an intermediate in the preparation of potassium persulfate by electrolysis. It is also employed in the manufacturing of fertilizers.

Solubility
Soluble in water, acetone and ethanol.

Notes
Hygroscopic. Incompatible with strong bases, strong oxidizing agents and alcohols.
TRUSTED_SUSTAINABILITY
Melting Point 195°C to 197°C (decomposition)
Quantity 100 g
Linear Formula KHSO4
UN Number UN2509
Merck Index 14,7613
Solubility Information Soluble in water,acetone and ethanol.
Formula Weight 136.17
Percent Purity 97%
Odor Odorless
Sensitivity Hygroscopic
Density 2.24
Chemical Name or Material Potassium hydrogen sulfate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'