missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium hexacyanoruthenate(II) hydrate, Ru 23.0% min, Thermo Scientific Chemicals
345.79 DKK valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
0.2 g
2 g
10 g
Description
Potassium hexacyanoruthenate(II) hydrate used in chemical research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | White |
Physical Form | Crystalline |
Quantity | 0.2 g |
Linear Formula | K4Ru(CN)6·xH2O |
UN Number | UN1588 |
Solubility Information | Soluble in water. |
Formula Weight | 413.57 (Anhydrous) |
Concentration or Composition (by Analyte or Components) | Ru 23.0% min. |
Chemical Name or Material | Potassium hexacyanoruthenate(II) hydrate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'