Få mere at vide
Potassium Dodecanoate-2,2-d2, CDN
Artikelnummer. 30321732
Skift visning
Mængde:
0.5 g
1 g
Pakningsstørrelse:
0.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30321732 0.5 g 0.5g
30315162 1 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30321732 Leverandør CDN Leverandørnr. CDND51670.5G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Potassium Dodecanoate-2,2-D2
CAS 82867-25-2
Molekylær formel C12 D2 H21 O2 . K
Mængde 0.5 g
Synonym Dodecanoic-2,2-d2 acid, potassium salt (9 CI), Potassium dodecanoate-2,2-d2, Potassium dodecanoate-2,2-D2, Potassium laurate-2,2-d2, Lauric-2,2-d2 acid potassium salt, Lauric acid-2,2-d2 potassium salt
InChI formel InChI=1 S/C12H24O2.K/c1-2-3-4-5-6-7-8-9-10-11-12(13)14;/h2-11H2,1H3,(H,13,14);/q;+1/p-1/i11D2;
SMIL [K+].[2 H]C([2 H])(CCCCCCCCCC)C(=O)[O-]
IUPAC navn potassium;2,2-dideuteriododecanoate
Molekylvægt (g/mol) 240.42
Formel vægt 240.146 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity