Learn More
Potassium Disulfate (Potassium Pyrosulfate), Supelco™
Product Code. 15834530
Change view
Click to view available options
Quantity:
1 kg
5 kg
Unit Size:
1kg
5kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15834530 5 kg 5kg
15824530 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15834530 Supplier Supelco Supplier No. 1051075000
 Restricted Item
Forventet forsendelse: 26-11-2026
For analysis EMSURE™ ACS
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 7790-62-7
Specifications
Melting Point 325°C
Quantity 5 kg
MDL Number MFCD00011385
Synonym Potassium disulfate (Potassium pyrosulfate), Potassium pyrosulfate, Potassium disulfate (Potassium pyrosulfate)
Molecular Weight (g/mol) 254.32
Grade Research
Chemical Name or Material Potassium Disulfate (Potassium Pyrosulfate)