Få mere at vide
Potassium Diisopropylxanthate, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Potassium Diisopropylxanthate |
| CAS | 3419-34-9 |
| Molekylær formel | C6H14KO2PS2 |
| Mængde | 1 g |
| Synonym | O,O-Diisopropyl Potassium Phosphorodithioate, Potassium O,O-Diisopropyl Dithiophosphate, Potassium O,O-Diisopropyl Phosphorodithioate, Potassium O,O'-Diisopropyl Dithiophosphate, Potassium Bis(O-isopropyl) Dithiophosphate, Potassium Diisopropyl Dithiophosphate |
| InChI formel | InChI=1S/C6H15O2PS2.K/c1-5(2)7-9(10,11)8-6(3)4;/h5-6H,1-4H3,(H,10,11);/q;+1/p-1 |
| SMIL | [K+].CC(C)OP(=S)([S-])OC(C)C |
| IUPAC navn | potassium;di(propan-2-yloxy)-sulfanylidene-sulfido-λ^{5}-phosphane |
| Molekylvægt (g/mol) | 252.38 |
| Formel vægt | 251.981 |